Assay Details
| Gene Symbol: Slc16a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857355.3 | XP_004857412.2 | 2 - 3 | 1232 | 103 | |
| XM_021238208.1 | XP_021093867.1 | 3 - 4 | 975 | 103 | ||
| XM_021238209.1 | XP_021093868.1 | 3 - 4 | 1136 | 103 | ||
| XM_021238210.1 | XP_021093869.1 | 3 - 4 | 989 | 103 | ||
| XM_021238211.1 | XP_021093870.1 | 4 - 5 | 1031 | 103 | ||
| XM_021238212.1 | XP_021093871.1 | 3 - 4 | 970 | 103 | ||
More Information
Set Membership: |
|

Slc16a11
Genomic Map