Assay Details
| Gene Symbol: Bsnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869639.3 | XP_004869696.1 | 1 - 2 | 420 | 59 | |
| XM_004869640.2 | XP_004869697.1 | 2 - 3 | 300 | 59 | ||
More Information
Set Membership: |
|

Bsnd
Genomic Map