Assay Details
| Gene Symbol: Bsnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869639.3 | XP_004869696.1 | 2 - 3 | 525 | 60 | |
| XM_004869640.2 | XP_004869697.1 | 3 - 4 | 405 | 60 | ||
More Information
Set Membership: |
|

Bsnd
Genomic Map