Assay Details
| Gene Symbol: Bsnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869639.3 | XP_004869696.1 | 3 - 4 | 782 | 92 | |
| XM_004869640.2 | XP_004869697.1 | 4 - 5 | 662 | 92 | ||
More Information
Set Membership: |
|

Bsnd
Genomic Map