Assay Details
| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873269.2 | XP_004873326.1 | 4 - 5 | 623 | 64 | |
| XM_021251108.1 | XP_021106767.1 | 5 - 6 | 786 | 64 | ||
| XM_021251109.1 | XP_021106768.1 | 4 - 5 | 557 | 64 | ||
More Information
Set Membership: |
|

Tmem38a
Genomic Map