Assay Details
| Gene Symbol: LOC101699488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242748.1 | XP_021098407.1 | 2 - 3 | 367 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101699488| Gene Symbol: LOC101699488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242748.1 | XP_021098407.1 | 2 - 3 | 367 | 73 | |
Set Membership: |
|