Assay Details
| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873790.3 | XP_004873847.1 | 6 - 7 | 633 | 66 | |
| XM_021251442.1 | XP_021107101.1 | 6 - 7 | 631 | 66 | ||
| XM_021251443.1 | XP_021107102.1 | 6 - 7 | 863 | 66 | ||
| XM_021251444.1 | XP_021107103.1 | 5 - 6 | 551 | 66 | ||
More Information
Set Membership: |
|

Rab40c
Genomic Map