Assay Details
| Gene Symbol: Znf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874872.2 | XP_004874929.1 | 1 - 2 | 534 | 82 | |
| XM_021252237.1 | XP_021107896.1 | 2 - 3 | 409 | 82 | ||
More Information
Set Membership: |
|

Znf74
Genomic Map