Assay Details
| Gene Symbol: Ripor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238957.1 | XP_021094616.1 | 18 - 19 | 3076 | 52 | |
| XM_021238958.1 | XP_021094617.1 | 18 - 19 | 3076 | 52 | ||
| XM_021238959.1 | XP_021094618.1 | 17 - 18 | 2953 | 52 | ||
| XM_021238960.1 | XP_021094619.1 | 17 - 18 | 2912 | 52 | ||
| XM_021238961.1 | XP_021094620.1 | 17 - 18 | 5868 | 52 | ||
| XM_021238962.1 | XP_021094621.1 | 17 - 18 | 3283 | 52 | ||
| XM_021238963.1 | XP_021094622.1 | 18 - 19 | 6015 | 52 | ||
More Information
Set Membership: |
|

Ripor3
Genomic Map