Assay Details
| Gene Symbol: LOC110347632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252646.1 | XP_021108305.1 | 2 - 3 | 193 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110347632| Gene Symbol: LOC110347632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252646.1 | XP_021108305.1 | 2 - 3 | 193 | 85 | |
Set Membership: |
|