Assay Details
| Gene Symbol: Snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852434.3 | XP_004852491.1 | 4 - 5 | 485 | 71 | |
| XM_013074980.2 | XP_012930434.1 | 4 - 5 | 488 | 71 | ||
| XM_021264606.1 | XP_021120265.1 | 3 - 4 | 459 | 71 | ||
More Information
Set Membership: |
|

Snx15
Genomic Map