Assay Details
| Gene Symbol: Rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859515.3 | XP_004859572.1 | 2 - 3 | 306 | 64 | |
| XM_021240003.1 | XP_021095662.1 | 2 - 3 | 249 | 64 | ||
More Information
Set Membership: |
|

Rpl19
Genomic Map