Assay Details
| Gene Symbol: Ttll6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077745.2 | XP_012933199.1 | 8 - 9 | 1573 | 60 | |
| XM_021240073.1 | XP_021095732.1 | 8 - 9 | 1574 | 60 | ||
| XM_021240074.1 | XP_021095733.1 | 8 - 9 | 1574 | 60 | ||
More Information
Set Membership: |
|

Ttll6
Genomic Map