Assay Details
| Gene Symbol: Tmem217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259902.1 | XP_021115561.1 | 1 - 2 | 408 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem217| Gene Symbol: Tmem217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259902.1 | XP_021115561.1 | 1 - 2 | 408 | 75 | |