Assay Details
| Gene Symbol: Rp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261112.1 | XP_021116771.1 | 1 - 2 | 62 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rp1l1| Gene Symbol: Rp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261112.1 | XP_021116771.1 | 1 - 2 | 62 | 100 | |