Assay Details
| Gene Symbol: Rp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261112.1 | XP_021116771.1 | 4 - 5 | 825 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rp1l1| Gene Symbol: Rp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261112.1 | XP_021116771.1 | 4 - 5 | 825 | 103 | |
Set Membership: |
|