Assay Details
| Gene Symbol: LOC101715932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263906.1 | XP_021119565.1 | 12 - 13 | 6654 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101715932| Gene Symbol: LOC101715932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263906.1 | XP_021119565.1 | 12 - 13 | 6654 | 70 | |
Set Membership: |
|