Assay Details
| Gene Symbol: LOC101715932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263906.1 | XP_021119565.1 | 1 - 2 | 2638 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021263906.1 transcripts.
Set Membership: |
|

LOC101715932
Genomic Map