Assay Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837642.2 | XP_004837699.1 | 5 - 6 | 1512 | 64 | |
| XM_021249846.1 | XP_021105505.1 | 3 - 4 | 627 | 64 | ||
| XM_021249855.1 | XP_021105514.1 | 4 - 5 | 695 | 64 | ||
| XM_021249860.1 | XP_021105519.1 | 4 - 5 | 718 | 64 | ||
More Information
Set Membership: |
|

Wnt5b
Genomic Map