Assay Details
| Gene Symbol: Dbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849590.2 | XP_004849647.1 | 9 - 10 | 1443 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbh| Gene Symbol: Dbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849590.2 | XP_004849647.1 | 9 - 10 | 1443 | 63 | |
Set Membership: |
|