Assay Details
| Gene Symbol: P3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859839.3 | XP_004859896.1 | 1 - 2 | 656 | 61 | |
| XM_021240331.1 | XP_021095990.1 | 1 - 2 | 661 | 61 | ||
| XM_021240332.1 | XP_021095991.1 | 1 - 2 | 658 | 61 | ||
| XM_021240333.1 | XP_021095992.1 | 1 - 2 | 659 | 61 | ||
| XM_021240334.1 | XP_021095993.1 | 1 - 2 | 666 | 61 | ||
More Information
Set Membership: |
|

P3h4
Genomic Map