Assay Details
| Gene Symbol: Gimap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860530.3 | XP_004860587.1 | 1 - 2 | 56 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004860530.3 transcripts.
Set Membership: |
|

Gimap4
Genomic Map