Assay Details
| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239885.1 | XP_021095544.1 | 6 - 7 | 1460 | 90 | |
| XM_021239886.1 | XP_021095545.1 | 6 - 7 | 1499 | 90 | ||
| XM_021239887.1 | XP_021095546.1 | 6 - 7 | 1410 | 90 | ||
| XM_021239888.1 | XP_021095547.1 | 6 - 7 | 1448 | 90 | ||
More Information
Set Membership: |
|

Wipf2
Genomic Map