Assay Details
| Gene Symbol: Cmtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843104.2 | XP_004843161.1 | 4 - 5 | 632 | 66 | |
| XM_021256938.1 | XP_021112597.1 | 4 - 5 | 841 | 66 | ||
| XM_021256939.1 | XP_021112598.1 | 4 - 5 | 678 | 66 | ||
| XM_021256940.1 | XP_021112599.1 | 4 - 5 | 618 | 66 | ||
More Information
Set Membership: |
|

Cmtm3
Genomic Map