Assay Details
| Gene Symbol: Bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851576.3 | XP_004851633.1 | 3 - 4 | 695 | 59 | |
| XM_021263942.1 | XP_021119601.1 | 4 - 5 | 915 | 59 | ||
More Information
Set Membership: |
|

Bbox1
Genomic Map