Assay Details
| Gene Symbol: Icmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863730.3 | XP_004863787.1 | 4 - 5 | 733 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Icmt| Gene Symbol: Icmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863730.3 | XP_004863787.1 | 4 - 5 | 733 | 68 | |
Set Membership: |
|