Assay Details
| Gene Symbol: Mmp24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246777.1 | XP_021102436.1 | 5 - 6 | 994 | 56 | |
| XM_021246778.1 | XP_021102437.1 | 6 - 7 | 1990 | 56 | ||
More Information
Set Membership: |
|

Mmp24
Genomic Map