Assay Details
| Gene Symbol: Llgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868692.3 | XP_004868749.1 | 15 - 16 | 2104 | 61 | |
| XM_021247591.1 | XP_021103250.1 | 15 - 16 | 2165 | 61 | ||
| XM_021247592.1 | XP_021103251.1 | 16 - 17 | 2138 | 61 | ||
| XM_021247593.1 | XP_021103252.1 | 15 - 16 | 2105 | 61 | ||
| XM_021247594.1 | XP_021103253.1 | 15 - 16 | 2164 | 61 | ||
More Information
Set Membership: |
|

Llgl1
Genomic Map