Assay Details
| Gene Symbol: Ninj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068701.2 | XP_012924155.1 | 2 - 3 | 307 | 72 | |
| XM_013068703.2 | XP_012924157.1 | 2 - 3 | 647 | 72 | ||
| XM_013068705.2 | XP_012924159.1 | 2 - 3 | 397 | 72 | ||
| XM_013068710.2 | XP_012924164.1 | 3 - 4 | 742 | 72 | ||
| XM_013068712.2 | XP_012924166.1 | 2 - 3 | 311 | 72 | ||
| XM_021249141.1 | XP_021104800.1 | 2 - 3 | 311 | 72 | ||
| XM_021249152.1 | XP_021104811.1 | 4 - 5 | 529 | 72 | ||
| XM_021249167.1 | XP_021104826.1 | 2 - 3 | 326 | 72 | ||
| XM_021249174.1 | XP_021104833.1 | 2 - 3 | 359 | 72 | ||
More Information
Set Membership: |
|

Ninj2
Genomic Map