Assay Details
| Gene Symbol: Slc35e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251132.1 | XP_021106791.1 | 6 - 7 | 976 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35e1| Gene Symbol: Slc35e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251132.1 | XP_021106791.1 | 6 - 7 | 976 | 61 | |
Set Membership: |
|