Assay Details
| Gene Symbol: Tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843723.3 | XP_004843780.1 | 2 - 3 | 1633 | 73 | |
| XM_021257394.1 | XP_021113053.1 | 2 - 3 | 1094 | 73 | ||
| XM_021257395.1 | XP_021113054.1 | 2 - 3 | 1345 | 73 | ||
| XM_021257396.1 | XP_021113055.1 | 3 - 4 | 726 | 73 | ||
| XM_021257397.1 | XP_021113056.1 | 1 - 2 | 1709 | 73 | ||
More Information
Set Membership: |
|

Tbx3
Genomic Map