Assay Details
| Gene Symbol: Shisa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004833936.3 | XP_004833993.1 | 5 - 6 | 721 | 64 | |
| XM_004833937.3 | XP_004833994.1 | 5 - 6 | 721 | 64 | ||
| XM_021257142.1 | XP_021112801.1 | 3 - 4 | 474 | 64 | ||
More Information
Set Membership: |
|

Shisa5
Genomic Map