Assay Details
| Gene Symbol: Coro1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843629.3 | XP_004843686.1 | 4 - 5 | 577 | 63 | |
| XM_013071715.2 | XP_012927169.1 | 3 - 4 | 472 | 63 | ||
More Information
Set Membership: |
|

Coro1c
Genomic Map