Assay Details
| Gene Symbol: LOC101715566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848494.3 | XP_004848551.1 | 1 - 2 | 424 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101715566| Gene Symbol: LOC101715566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848494.3 | XP_004848551.1 | 1 - 2 | 424 | 103 | |
Set Membership: |
|