Assay Details
| Gene Symbol: Sgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858206.3 | XP_004858263.1 | 12 - 13 | 1150 | 66 | |
| XM_004858212.3 | XP_004858269.1 | 12 - 13 | 1132 | 66 | ||
More Information
Set Membership: |
|

Sgk2
Genomic Map