Assay Details
| Gene Symbol: Mrps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859808.2 | XP_004859865.1 | 2 - 3 | 313 | 86 | |
| XM_021240130.1 | XP_021095789.1 | 2 - 3 | 353 | 86 | ||
| XM_021240131.1 | XP_021095790.1 | 2 - 3 | 353 | 86 | ||
| XM_021240132.1 | XP_021095791.1 | 2 - 3 | 353 | 86 | ||
More Information
Set Membership: |
|

Mrps23
Genomic Map