Assay Details
| Gene Symbol: Znf444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866601.3 | XP_004866658.1 | 2 - 3 | 207 | 88 | |
| XM_004866602.3 | XP_004866659.1 | 2 - 3 | 222 | 88 | ||
| XM_021246253.1 | XP_021101912.1 | 2 - 3 | 191 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004866601.3, XM_004866602.3, XM_021246253.1 transcripts.
Set Membership: |
|

Znf444
Genomic Map