Assay Details
| Gene Symbol: Znf444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866601.3 | XP_004866658.1 | 3 - 4 | 574 | 63 | |
| XM_004866602.3 | XP_004866659.1 | 3 - 4 | 589 | 63 | ||
| XM_021246253.1 | XP_021101912.1 | 3 - 4 | 558 | 63 | ||
More Information
Set Membership: |
|

Znf444
Genomic Map