Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867077.3 | XP_004867134.1 | 12 - 13 | 2159 | 59 | |
| XM_004867079.3 | XP_004867136.1 | 13 - 14 | 2179 | 59 | ||
| XM_004867080.3 | XP_004867137.1 | 12 - 13 | 2159 | 59 | ||
| XM_004867081.3 | XP_004867138.1 | 12 - 13 | 2117 | 59 | ||
| XM_013066347.2 | XP_012921801.1 | 12 - 13 | 2060 | 59 | ||
| XM_013066348.2 | XP_012921802.1 | 13 - 14 | 2070 | 59 | ||
| XM_021245744.1 | XP_021101403.1 | 12 - 13 | 2036 | 59 | ||
| XM_021245745.1 | XP_021101404.1 | 12 - 13 | 2089 | 59 | ||
More Information
Set Membership: |
|

Zc3h4
Genomic Map