Assay Details
| Gene Symbol: Runx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846437.3 | XP_004846494.1 | 4 - 5 | 979 | 87 | |
| XM_013072895.2 | XP_012928349.1 | 4 - 5 | 1611 | 87 | ||
| XM_021259723.1 | XP_021115382.1 | 4 - 5 | 979 | 87 | ||
| XM_021259724.1 | XP_021115383.1 | 4 - 5 | 977 | 87 | ||
| XM_021259725.1 | XP_021115384.1 | 3 - 4 | 731 | 87 | ||
| XM_021259726.1 | XP_021115385.1 | 4 - 5 | 977 | 87 | ||
| XM_021259727.1 | XP_021115386.1 | 4 - 5 | 981 | 87 | ||
| XM_021259728.1 | XP_021115387.1 | 2 - 3 | 529 | 87 | ||
More Information
Set Membership: |
|

Runx2
Genomic Map