Assay Details
| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848335.2 | XP_004848392.1 | 8 - 9 | 2519 | 50 | |
| XM_004848336.3 | XP_004848393.1 | 7 - 8 | 2352 | 50 | ||
More Information
Set Membership: |
|

Rhobtb2
Genomic Map