Assay Details
| Gene Symbol: Dsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853123.2 | XP_004853180.1 | 12 - 13 | 2009 | 58 | |
| XM_021265120.1 | XP_021120779.1 | 11 - 12 | 1862 | 58 | ||
More Information
Set Membership: |
|

Dsg2
Genomic Map