Assay Details
| Gene Symbol: Kctd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854340.2 | XP_004854397.1 | 4 - 5 | 1552 | 68 | |
| XM_021266153.1 | XP_021121812.1 | 4 - 5 | 1498 | 68 | ||
More Information
Set Membership: |
|

Kctd16
Genomic Map