Assay Details
| Gene Symbol: Gpr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247035.1 | XP_021102694.1 | 2 - 3 | 420 | 88 | |
| XM_021247047.1 | XP_021102706.1 | 3 - 4 | 1264 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021247047.1 transcripts.
Set Membership: |
|

Gpr55
Genomic Map