Assay Details
| Gene Symbol: LOC110343890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247832.1 | XP_021103491.1 | 1 - 2 | 261 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110343890| Gene Symbol: LOC110343890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247832.1 | XP_021103491.1 | 1 - 2 | 261 | 65 | |
Set Membership: |
|