Assay Details
| Gene Symbol: Fkbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840073.2 | XP_004840130.1 | 8 - 9 | 1086 | 72 | |
| XM_021254422.1 | XP_021110081.1 | 9 - 10 | 1201 | 72 | ||
| XM_021254423.1 | XP_021110082.1 | 8 - 9 | 1056 | 72 | ||
| XM_021254424.1 | XP_021110083.1 | 7 - 8 | 996 | 72 | ||
More Information
Set Membership: |
|

Fkbp6
Genomic Map