Assay Details
| Gene Symbol: Gjb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854878.2 | XP_004854935.1 | 3 - 4 | 573 | 81 | |
| XM_004854879.3 | XP_004854936.1 | 2 - 3 | 675 | 81 | ||
| XM_004854880.2 | XP_004854937.1 | 3 - 4 | 950 | 81 | ||
| XM_004854881.2 | XP_004854938.1 | 2 - 3 | 868 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004854878.2, XM_004854879.3, XM_004854880.2, XM_004854881.2 transcripts.
Set Membership: |
|

Gjb6
Genomic Map