Assay Details
| Gene Symbol: Fcrla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021244704.1 | XP_021100363.1 | 2 - 3 | 572 | 59 | |
| XM_021244705.1 | XP_021100364.1 | 2 - 3 | 288 | 59 | ||
More Information
Set Membership: |
|

Fcrla
Genomic Map