Assay Details
| Gene Symbol: Cdcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835048.2 | XP_004835105.1 | 7 - 8 | 2032 | 65 | |
| XM_021257268.1 | XP_021112927.1 | 7 - 8 | 2032 | 65 | ||
| XM_021257274.1 | XP_021112933.1 | 7 - 8 | 2266 | 65 | ||
More Information
Set Membership: |
|

Cdcp1
Genomic Map