Assay Details
Gene Symbol: LOC101699174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843301.3 | XP_004843358.1 | 2 - 3 | 568 | 100 | |
XM_004843302.3 | XP_004843359.1 | 2 - 3 | 570 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
